Assay Details
| Gene Symbol: C11H17orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597224.3 | XP_005597281.1 | 1898 | 70 | ||
| XM_005597225.3 | XP_005597282.1 | 1798 | 70 | |||
| XM_005597226.3 | XP_005597283.1 | 1897 | 70 | |||
| XM_005597227.3 | XP_005597284.1 | 1897 | 70 | |||
| XM_014739026.2 | XP_014594512.1 | 1857 | 70 | |||
| XM_023652117.1 | XP_023507885.1 | 1745 | 70 | |||
More Information
Set Membership: |
|

C11H17orf80
Genomic Map