Assay Details
| Gene Symbol: EIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362563.4 | XP_003362611.1 | 175 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003362563.4 transcripts.
Set Membership: |
|

EIF1
Genomic Map