Assay Details
| Gene Symbol: KRT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597407.3 | XP_005597464.1 | 1154 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT15| Gene Symbol: KRT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597407.3 | XP_005597464.1 | 1154 | 92 | ||