Assay Details
Gene Symbol: LOC102148710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597558.3 | XP_005597615.1 | 275 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
