Assay Details
| Gene Symbol: LOC100058771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597602.3 | XP_005597659.1 | 139 | 79 | ||
| XM_005597603.3 | XP_005597660.1 | 138 | 79 | |||
| XM_005597604.3 | XP_005597661.1 | 139 | 79 | |||
| XM_005597605.3 | XP_005597662.1 | 138 | 79 | |||
| XM_005597606.3 | XP_005597663.1 | 138 | 79 | |||
| XM_005597607.3 | XP_005597664.1 | 138 | 79 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005597602.3, XM_005597603.3, XM_005597604.3, XM_005597605.3, XM_005597606.3, XM_005597607.3 transcripts.
Set Membership: |
|

LOC100058771
Genomic Map