Assay Details
| Gene Symbol: PIMREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504737.4 | XP_001504787.1 | 732 | 68 | ||
| XM_005597686.3 | XP_005597743.1 | 732 | 68 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIMREG| Gene Symbol: PIMREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504737.4 | XP_001504787.1 | 732 | 68 | ||
| XM_005597686.3 | XP_005597743.1 | 732 | 68 | |||
Set Membership: |
|