Assay Details
| Gene Symbol: CXCL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504756.6 | XP_001504806.1 | 658 | 95 | ||
| XM_023653383.1 | XP_023509151.1 | 525 | 95 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXCL16| Gene Symbol: CXCL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504756.6 | XP_001504806.1 | 658 | 95 | ||
| XM_023653383.1 | XP_023509151.1 | 525 | 95 | |||
Set Membership: |
|