Assay Details
| Gene Symbol: LOC100072916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014737319.2 | XP_014592805.2 | 1299 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100072916| Gene Symbol: LOC100072916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014737319.2 | XP_014592805.2 | 1299 | 75 | ||
Set Membership: |
|