Assay Details
| Gene Symbol: RPL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504821.5 | XP_001504871.1 | 474 | 66 | ||
| XM_014728693.1 | XP_014584179.1 | 339 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL26| Gene Symbol: RPL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504821.5 | XP_001504871.1 | 474 | 66 | ||
| XM_014728693.1 | XP_014584179.1 | 339 | 66 | |||
Set Membership: |
|