Assay Details
| Gene Symbol: CHST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023653927.1 | XP_023509695.1 | 1019 | 87 | ||
| XM_023653928.1 | XP_023509696.1 | 790 | 87 | |||
| XM_023653929.1 | XP_023509697.1 | 956 | 87 | |||
| XM_023653930.1 | XP_023509698.1 | 904 | 87 | |||
| XM_023653931.1 | XP_023509699.1 | 809 | 87 | |||
| XM_023653932.1 | XP_023509700.1 | 695 | 87 | |||
| XM_023653933.1 | XP_023509701.1 | 346 | 87 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023653927.1, XM_023653928.1, XM_023653929.1, XM_023653930.1, XM_023653931.1, XM_023653932.1, XM_023653933.1 transcripts.
Set Membership: |
|

CHST1
Genomic Map