Assay Details
| Gene Symbol: FAM111A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598161.3 | XP_005598218.1 | 840 | 145 | ||
| XM_023654100.1 | XP_023509868.1 | 867 | 145 | |||
| XM_023654101.1 | XP_023509869.1 | 825 | 145 | |||
| GenBank mRNA | JL629938.1 | - | 1 - 2 | 128 | 144 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005598161.3, XM_023654100.1, XM_023654101.1 transcripts.
Set Membership: |
|

FAM111A
Genomic Map