Assay Details
| Gene Symbol: PLAAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598258.3 | XP_005598315.2 | 570 | 81 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLAAT5| Gene Symbol: PLAAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598258.3 | XP_005598315.2 | 570 | 81 | ||