Assay Details
| Gene Symbol: IFI44L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496470.6 | XP_001496520.4 | 127 | 90 | ||
| XM_005610477.3 | XP_005610534.2 | 127 | 90 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001496470.6, XM_005610477.3 transcripts.
Set Membership: |
|

IFI44L
Genomic Map