Assay Details
| Gene Symbol: GAL3ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495167.5 | XP_001495217.1 | 225 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001495167.5 transcripts.
Set Membership: |
|

GAL3ST3
Genomic Map