Assay Details
| Gene Symbol: CDK2AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001917559.4 | XP_001917594.1 | 753 | 141 | ||
| XM_005598345.3 | XP_005598402.1 | 890 | 283 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005598345.3 transcripts.
Set Membership: |
|

CDK2AP2
Genomic Map