Assay Details
| Gene Symbol: CDK2AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001917559.4 | XP_001917594.1 | 892 | 94 | ||
| XM_005598345.3 | XP_005598402.1 | 1029 | 94 | |||
More Information
Set Membership: |
|

CDK2AP2
Genomic Map