Assay Details
| Gene Symbol: LOC102148181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729771.2 | XP_014585257.1 | 5122 | 55 | ||
| XM_023654831.1 | XP_023510599.1 | 5228 | 55 | |||
| XM_023654832.1 | XP_023510600.1 | 10593 | 55 | |||
| XM_023654833.1 | XP_023510601.1 | 6165 | 55 | |||
| XM_023654834.1 | XP_023510602.1 | 5261 | 55 | |||
| XM_023654835.1 | XP_023510603.1 | 5264 | 55 | |||
| XM_023654836.1 | XP_023510604.1 | 2550 | 55 | |||
| XM_023654837.1 | XP_023510605.1 | 3054 | 55 | |||
| XM_023654838.1 | XP_023510606.1 | 2708 | 55 | |||
| XM_023654840.1 | XP_023510608.1 | 5142 | 55 | |||
| XM_023654841.1 | XP_023510609.1 | 4382 | 55 | |||
| XM_023654842.1 | XP_023510610.1 | 925 | 55 | |||
| XM_023654843.1 | XP_023510611.1 | 6858 | 55 | |||
| XM_023654844.1 | XP_023510612.1 | 6819 | 55 | |||
| XM_023654845.1 | XP_023510613.1 | 5036 | 55 | |||
More Information
Set Membership: |
|

LOC102148181
Genomic Map