Assay Details
| Gene Symbol: LOC100059180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655251.1 | XP_023511019.1 | 2409 | 92 | ||
| XM_023655253.1 | XP_023511021.1 | 2265 | 92 | |||
More Information
Set Membership: |
|

LOC100059180
Genomic Map