Assay Details
| Gene Symbol: MAFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655107.1 | XP_023510875.1 | 1741 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023655107.1 transcripts.
Set Membership: |
|

MAFK
Genomic Map