Assay Details
| Gene Symbol: MAFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655107.1 | XP_023510875.1 | 1885 | 64 | ||
| XM_023655108.1 | XP_023510876.1 | 327 | 64 | |||
| XM_023655109.1 | XP_023510877.1 | 573 | 64 | |||
| XM_023655110.1 | XP_023510878.1 | 708 | 64 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023655107.1, XM_023655108.1, XM_023655109.1, XM_023655110.1 transcripts.
Set Membership: |
|

MAFK
Genomic Map