Assay Details
| Gene Symbol: POM121C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615124.2 | XP_005615181.2 | 1331 | 94 | ||
| XM_023655427.1 | XP_023511195.1 | 1250 | 94 | |||
| XM_023655428.1 | XP_023511196.1 | 1016 | 94 | |||
| XM_023655429.1 | XP_023511197.1 | 859 | 94 | |||
More Information
Set Membership: |
|

POM121C
Genomic Map