Assay Details
Gene Symbol: LOC100062625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494135.4 | XP_001494185.1 | 1588 | 104 | ||
XM_023655502.1 | XP_023511270.1 | 1646 | 104 | |||
XM_023655503.1 | XP_023511271.1 | 1620 | 104 | |||
XM_023655504.1 | XP_023511272.1 | 1569 | 104 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
