Assay Details
Gene Symbol: LOC100062625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494135.4 | XP_001494185.1 | 609 | 106 | ||
XM_023655502.1 | XP_023511270.1 | 667 | 106 | |||
XM_023655503.1 | XP_023511271.1 | 668 | 106 | |||
XM_023655504.1 | XP_023511272.1 | 668 | 106 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
