Assay Details
| Gene Symbol: LOC100062082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598712.3 | XP_005598769.2 | 1720 | 67 | ||
| XM_014729938.2 | XP_014585424.2 | 1771 | 67 | |||
| XM_023655521.1 | XP_023511289.1 | 1772 | 67 | |||
| XM_023655522.1 | XP_023511290.1 | 1720 | 67 | |||
| XM_023655523.1 | XP_023511291.1 | 1578 | 67 | |||
| XM_023655524.1 | XP_023511292.1 | 1773 | 67 | |||
| XM_023655525.1 | XP_023511293.1 | 1731 | 67 | |||
| GenBank mRNA | JL619270.1 | - | 1 - 2 | 176 | 67 | |
More Information
Set Membership: |
|

LOC100062082
Genomic Map