Assay Details
| Gene Symbol: LOC100062082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598712.3 | XP_005598769.2 | 1859 | 98 | ||
| XM_014729938.2 | XP_014585424.2 | 1910 | 98 | |||
| GenBank mRNA | JL619270.1 | - | 2 - 3 | 315 | 98 | |
More Information
Set Membership: |
|

LOC100062082
Genomic Map