Assay Details
| Gene Symbol: LOC100062082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598712.3 | XP_005598769.2 | 2127 | 82 | ||
| XM_014729938.2 | XP_014585424.2 | 2178 | 82 | |||
| XM_023655521.1 | XP_023511289.1 | 2215 | 82 | |||
| XM_023655522.1 | XP_023511290.1 | 2163 | 82 | |||
| XM_023655523.1 | XP_023511291.1 | 2021 | 82 | |||
| XM_023655525.1 | XP_023511293.1 | 2174 | 82 | |||
| GenBank mRNA | JL619270.1 | - | 4 - 5 | 583 | 82 | |
More Information
Set Membership: |
|

LOC100062082
Genomic Map