Assay Details
| Gene Symbol: C13H16orf92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362740.4 | XP_003362788.1 | 343 | 65 | ||
| XM_023616033.1 | XP_023471801.1 | 346 | 65 | |||
More Information
Set Membership: |
|

C13H16orf92
Genomic Map