Assay Details
| Gene Symbol: LOC100058833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023616174.1 | XP_023471942.1 | 3949 | 132 | ||
| XM_023616175.1 | XP_023471943.1 | 3743 | 132 | |||
| XM_023616177.1 | XP_023471945.1 | 3949 | 132 | |||
| XM_023616178.1 | XP_023471946.1 | 3850 | 132 | |||
| XM_023616179.1 | XP_023471947.1 | 3795 | 132 | |||
| XM_023616180.1 | XP_023471948.1 | 3571 | 132 | |||
More Information
Set Membership: |
|

LOC100058833
Genomic Map