Assay Details
| Gene Symbol: LOC100058987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001491761.5 | XP_001491811.3 | 153 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001491761.5 transcripts.
Set Membership: |
|

LOC100058987
Genomic Map