Assay Details
| Gene Symbol: LOC100058987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001491761.5 | XP_001491811.3 | 3580 | 84 | ||
| XM_023616173.1 | XP_023471941.1 | 3450 | 84 | |||
More Information
Set Membership: |
|

LOC100058987
Genomic Map