Assay Details
| Gene Symbol: CEP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362733.3 | XP_003362781.1 | 385 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEP20| Gene Symbol: CEP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362733.3 | XP_003362781.1 | 385 | 80 | ||