Assay Details
| Gene Symbol: MRTFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598901.3 | XP_005598958.1 | 7647 | 83 | ||
| XM_023616297.1 | XP_023472065.1 | 4184 | 83 | |||
| XM_023616298.1 | XP_023472066.1 | 4197 | 83 | |||
| XM_023616299.1 | XP_023472067.1 | 2886 | 83 | |||
| XM_023616300.1 | XP_023472068.1 | 2823 | 83 | |||
| XM_023616301.1 | XP_023472069.1 | 4051 | 83 | |||
More Information
Set Membership: |
|

MRTFB
Genomic Map