Assay Details
| Gene Symbol: SEPTIN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598989.3 | XP_005599046.2 | 933 | 71 | ||
| XM_023616374.1 | XP_023472142.1 | 975 | 71 | |||
| XM_023616375.1 | XP_023472143.1 | 948 | 71 | |||
| XM_023616376.1 | XP_023472144.1 | 822 | 71 | |||
| XM_023616377.1 | XP_023472145.1 | 975 | 71 | |||
More Information
Set Membership: |
|

SEPTIN12
Genomic Map