Assay Details
| Gene Symbol: LOC100069750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023616409.1 | XP_023472177.1 | 811 | 88 | ||
| XM_023616410.1 | XP_023472178.1 | 367 | 88 | |||
| XM_023616411.1 | XP_023472179.1 | 812 | 88 | |||
| XM_023616412.1 | XP_023472180.1 | 168 | 88 | |||
More Information
Set Membership: |
|

LOC100069750
Genomic Map