Assay Details
| Gene Symbol: LOC100068977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599036.3 | XP_005599093.3 | 2263 | 91 | ||
| XM_023616456.1 | XP_023472224.1 | 2263 | 91 | |||
| XM_023616457.1 | XP_023472225.1 | 2179 | 91 | |||
| XM_023616459.1 | XP_023472227.1 | 2095 | 91 | |||
| XM_023616460.1 | XP_023472228.1 | 2263 | 91 | |||
More Information
Set Membership: |
|

LOC100068977
Genomic Map