Assay Details
| Gene Symbol: IL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599041.3 | XP_005599098.1 | 164 | 66 | ||
| XM_005599042.3 | XP_005599099.1 | 253 | 66 | |||
| XM_014730222.2 | XP_014585708.1 | 166 | 66 | |||
| XM_014730223.2 | XP_014585709.1 | 259 | 66 | |||
| XM_023616471.1 | XP_023472239.1 | 253 | 66 | |||
| XM_023616472.1 | XP_023472240.1 | 259 | 66 | |||
| XM_023616474.1 | XP_023472242.1 | 258 | 66 | |||
More Information
Set Membership: |
|

IL32
Genomic Map