Assay Details
| Gene Symbol: IL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599041.3 | XP_005599098.1 | 217 | 101 | ||
| XM_023616474.1 | XP_023472242.1 | 311 | 101 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: IL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599041.3 | XP_005599098.1 | 217 | 101 | ||
| XM_023616474.1 | XP_023472242.1 | 311 | 101 | |||
Set Membership: |
|