Assay Details
| Gene Symbol: IL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599041.3 | XP_005599098.1 | 242 | 84 | ||
| XM_014730222.2 | XP_014585708.1 | 334 | 174 | |||
| XM_014730223.2 | XP_014585709.1 | 427 | 174 | |||
| XM_023616474.1 | XP_023472242.1 | 336 | 84 | |||
More Information
Set Membership: |
|

IL32
Genomic Map