Assay Details
| Gene Symbol: IL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599041.3 | XP_005599098.1 | 300 | 67 | ||
| XM_005599042.3 | XP_005599099.1 | 362 | 67 | |||
| XM_014730222.2 | XP_014585708.1 | 392 | 67 | |||
| XM_023616471.1 | XP_023472239.1 | 452 | 67 | |||
More Information
Set Membership: |
|

IL32
Genomic Map