Assay Details
| Gene Symbol: CCDC154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599072.3 | XP_005599129.1 | 2293 | 109 | ||
| XM_023616658.1 | XP_023472426.1 | 2508 | 109 | |||
More Information
Set Membership: |
|

CCDC154
Genomic Map