Assay Details
| Gene Symbol: LOC100067178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599073.3 | XP_005599130.1 | 1291 | 84 | ||
| XM_014730268.2 | XP_014585754.1 | 1279 | 84 | |||
| XM_023616684.1 | XP_023472452.1 | 1357 | 84 | |||
More Information
Set Membership: |
|

LOC100067178
Genomic Map