Assay Details
| Gene Symbol: CIAO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496246.4 | XP_001496296.1 | 916 | 61 | ||
| XM_014730269.2 | XP_014585755.1 | 916 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CIAO3| Gene Symbol: CIAO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496246.4 | XP_001496296.1 | 916 | 61 | ||
| XM_014730269.2 | XP_014585755.1 | 916 | 61 | |||
Set Membership: |
|