Assay Details
| Gene Symbol: CIAO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496246.4 | XP_001496296.1 | 990 | 100 | ||
| XM_023616685.1 | XP_023472453.1 | 1023 | 100 | |||
More Information
Set Membership: |
|

CIAO3
Genomic Map