Assay Details
| Gene Symbol: PTTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503444.5 | XP_001503494.1 | 524 | 77 | ||
| XM_005599239.3 | XP_005599296.1 | 486 | 77 | |||
| XM_005599240.3 | XP_005599297.1 | 459 | 77 | |||
| XM_005599241.3 | XP_005599298.1 | 515 | 77 | |||
| XM_005599243.3 | XP_005599300.1 | 513 | 77 | |||
| XM_023617294.1 | XP_023473062.1 | 458 | 77 | |||
More Information
Set Membership: |
|

PTTG1
Genomic Map