Assay Details
| Gene Symbol: CCNJL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599251.3 | XP_005599308.2 | 896 | 84 | ||
| XM_023617300.1 | XP_023473068.1 | 928 | 84 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCNJL| Gene Symbol: CCNJL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599251.3 | XP_005599308.2 | 896 | 84 | ||
| XM_023617300.1 | XP_023473068.1 | 928 | 84 | |||
Set Membership: |
|