Assay Details
| Gene Symbol: C14H5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504379.5 | XP_001504429.1 | 267 | 84 | ||
| XM_005599410.3 | XP_005599467.1 | 266 | 84 | |||
| XM_023617675.1 | XP_023473443.1 | 267 | 84 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001504379.5, XM_005599410.3, XM_023617675.1 transcripts.
Set Membership: |
|

C14H5orf24
Genomic Map