Assay Details
| Gene Symbol: LOC100073089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596931.3 | XP_005596988.2 | 1752 | 111 | ||
| XM_005596932.3 | XP_005596989.1 | 1402 | 111 | |||
| XM_023651094.1 | XP_023506862.1 | 1891 | 111 | |||
| XM_023651095.1 | XP_023506863.1 | 1888 | 111 | |||
| XM_023651096.1 | XP_023506864.1 | 1891 | 111 | |||
| XM_023651097.1 | XP_023506865.1 | 1891 | 111 | |||
| XM_023651098.1 | XP_023506866.1 | 1891 | 111 | |||
More Information
Set Membership: |
|

LOC100073089
Genomic Map