Assay Details
| Gene Symbol: LOC100054688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501646.5 | XP_001501696.3 | 750 | 76 | ||
| XM_005600091.3 | XP_005600148.2 | 754 | 76 | |||
| XM_005600092.3 | XP_005600149.2 | 754 | 76 | |||
| XM_014731220.2 | XP_014586706.2 | 752 | 76 | |||
| XM_023619280.1 | XP_023475048.1 | 754 | 76 | |||
More Information
Set Membership: |
|

LOC100054688
Genomic Map