Assay Details
| Gene Symbol: LOC100147344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625343.1 | XP_023481111.1 | 1285 | 115 | ||
| XM_023625344.1 | XP_023481112.1 | 1285 | 115 | |||
| XM_023625345.1 | XP_023481113.1 | 1365 | 115 | |||
| XM_023625346.1 | XP_023481114.1 | 1364 | 115 | |||
| XM_023625347.1 | XP_023481115.1 | 2118 | 115 | |||
| XM_023625348.1 | XP_023481116.1 | 1455 | 115 | |||
More Information
Set Membership: |
|

LOC100147344
Genomic Map