Assay Details
| Gene Symbol: SPAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023639591.1 | XP_023495359.1 | 2436 | 157 | ||
| XM_023639592.1 | XP_023495360.1 | 2529 | 157 | |||
| XM_023639593.1 | XP_023495361.1 | 2468 | 157 | |||
| XM_023639594.1 | XP_023495362.1 | 2543 | 157 | |||
| XM_023639595.1 | XP_023495363.1 | 2658 | 157 | |||
More Information
Set Membership: |
|

SPAM1
Genomic Map